missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Varicella Zoster Virus VZV ORF9 Protein

A cDNA sequence encoding the VZV ORF9 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13952-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985219

Quantity Price
1 757.92 € / 500µg
5 711.76 € / 500µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

Varicella Zoster Virus VZV ORF9 Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Varicella Zoster Virus
Untagged
Varicella protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
25mM Tris-Hcl pH 8.0, 1mM EDTA and 50% glycerol.
Videos
SDS
Documents
Certificates