missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus Norovirus Group-2 Protein
A cDNA sequence encoding the Norovirus Group-2 was constructed and used to recombinantly synthesize the protein.
228.00 € - 1740.00 €
Specifications
Name | Virus Norovirus Group-2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15991789
|
enQuireBio™
QP12867-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911799
|
enQuireBio™
QP12867-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15901799
|
enQuireBio™
QP12867-1MG |
1 mg |
1740.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Virus Norovirus Group-2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Virus | |
His | |
Phosphate buffer, 1M urea and 50mM arginine. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKMASNDATP SNDGAAGLVP EINNEAMALD PVAGAAIAAP LTGQQNIIDP WIMNNFVQAP GGEFTVSPRN SPGEVLLNLE LGPEINPYLA HLARMYNGYA GGFEVQVVLA GNAFTAGKII FAAIPPNFPI ENLSAAQITM CPHVIVDVRQ LEPVNLPMPD VRNNFFHYNQ GSDSKLRLVA MLYTPLRANN SGDDVFTVSC RVLTRPSPDF SFNFLVPPTV ESKTKPFTLP ILTISEMSNS RFPVPIDSLH TSPTENIVVQ CQNGRVTLDG ELMGTTQLLP SQICAFRGVL TRSTSRASDQ ADTATPRLFN YYWHIQLDNL NGTPYDPAED IPGPLGTPDF RGKVFGVASQ RNPDSTTRAH EAKVDTTAGR FTPKLGSLEI STESGDFDQN QPTRFTPVGI GVDHEADFQQ WSLPDYSGQF THNMNLAPAV APNFPGEQLL FFRSQLPSSG GRSNGILDCR VPQEWVQHFY QESAPAQTQV ALVRYVNPDT GRVLFEAKLH KLGFMTIAKN GDSPITVPPN GYFRFESWVN PFYTLAPMGT GNGRRRIQ | |
Protein is >85% pure as determined by 12% PAGE (coomassie staining). |