missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus VEGF E (Orf Virus) Protein
A cDNA sequence encoding the VEGF E (Orf Virus) was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10912-2ug
Additional Details : Weight : 0.01000kg
Specifications
Virus VEGF E (Orf Virus) Protein | |
Research Use Only | |
The biological activity was determined (1) by the ability to induce VEGFR-2/KDR receptor phosphorylation in PAE/KDR cells and (2) in a cell proliferation assay using primary HUVECs. The ED50 for this effect is typically 1-5ng/ml. | |
Virus | |
His | |
The protein was lyophilized from a concentrated (1 mg/ml) solution containing PBS. |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MKLLVGILVA VCLHQYLLNA DSNTKGWSEV LKGSECKPRP IVVPVSETHP ELTSQRFNPP CVTLMRCGGC CNDESLECVP TEEVNVTMEL LGASGSGSNG MQRLSFVEHK KCDCRPRFTT TPPTTTRPPR RRR | |
Greater than 90.0% as determined by SDS-PAGE. |