missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Yeast GLRX1 Yeast Protein
A cDNA sequence encoding the GLRX1 Yeast was constructed and used to recombinantly synthesize the protein.
131.00 € - 2175.00 €
Specifications
Gene ID (Entrez) | 850322 |
---|---|
Name | GLRX1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | GRX1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15989018
|
enQuireBio™
QP11990-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15909028
|
enQuireBio™
QP11990-50UG |
50 μg |
203.00 €
50µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999018
|
enQuireBio™
QP11990-1MG |
1 mg |
2175.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
850322 | |
Research Use Only | |
GRX1 | |
Yeast | |
His | |
Glutaredoxin solution contains 25mM Tris-HCl pH 7.5 and 0.01% Na Azide. |
GLRX1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MVSQETIKHV KDLIAENEIF VASKTYCPYC HAALNTLFEK LKVPRSKVLV LQLNDMKEGA DIQAALYEIN GQRTVPNIYI NGKHIGGNDD LQELRETGEL EELLEPILAN | |
Greater than 90% as determined by SDS-PAGE. |