missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RedTaq DNA Polymerase
RedTaq DNA polymerase catalyzes 5'-3' synthesis of DNA.
Brand: Abnova™ D0012.2500Units
Additional Details : Weight : 0.02000kg
Description
- The enzyme has been proved not having the 3′-5′ exonuclease activity.
- The enzyme has proven to have high amplification yield, be stable at high temparetaure.
- Added inert dye will not have any interference to the reaction.
- Visual confirmation that the enzyme has been added and proper component mixing of the reaction has occurred.
- Samples can be loaded directly onto an agarose gel for electrophoresis with no loading dye addition.
Specifications
Store at -20°C Guaranteed stable for 6 months when properly stored | |
Standard PCR | |
20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol |
2500 units | |
1 unit/μL |