missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RedTaq DNA Polymerase
RedTaq DNA polymerase catalyzes 5'-3' synthesis of DNA.
195.00 € - 755.00 €
Specifications
Content And Storage | Store at -20°C Guaranteed stable for 6 months when properly stored |
---|---|
For Use With (Application) | Standard PCR |
Concentration | 1 unit/μL |
Storage Buffer | 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022555
|
Abnova™
D0012.1000UNITS |
1000 units |
378.00 €
1000 units |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032555
|
Abnova™
D0012.2500UNITS |
2500 units |
755.00 €
2500 units |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042555
|
Abnova™
D0012.500UNITS |
500 units |
195.00 €
500 units |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- The enzyme has been proved not having the 3′-5′ exonuclease activity.
- The enzyme has proven to have high amplification yield, be stable at high temparetaure.
- Added inert dye will not have any interference to the reaction.
- Visual confirmation that the enzyme has been added and proper component mixing of the reaction has occurred.
- Samples can be loaded directly onto an agarose gel for electrophoresis with no loading dye addition.
Specifications
Store at -20°C Guaranteed stable for 6 months when properly stored | |
Standard PCR | |
1 unit/μL | |
20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol |