Learn More
Reichert™ Rhino Ultra-Durable Handheld Automotive Fluids Refractometers, DC60 Duo-Che; Celsius
Brand: Reichert™ 81000-05
Code : A0
Additional Details : Weight : 0.80000kg
Description
Duo-Chek models test engine coolant and battery charge in factory and field use—fully IP67 water rresistant, dustproof, and impact resistant
Order Info
Rhino Model for Industrial Fluid Testing is also available; see Cat. No. RE-10440.