missing translation for 'onlineSavingsMsg'
Learn More

Reinecke Salt, ACS Reagent, Thermo Scientific Chemicals

Catalog Number p-3734724
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13573-16-5
C4H10CrN7S4·H2O
354.46
MFCD00150925
Ammonium reineckate, Ammonium tetrathiocyanatodiamminechromate
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13573-16-5
C4H10CrN7S4·H2O
354.46
MFCD00150925
Ammonium reineckate, Ammonium tetrathiocyanatodiamminechromate

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 270.0°C
Color Red
Physical Form Crystalline Powder
Quantity 25 g
Linear Formula NH4(Cr(NH3)2(SCN)4)·H2O
Merck Index 15, 8248
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in deluted hydrochloric acid,soluble in ethanol
Formula Weight 354.46
Grade ACS Reagent
Infrared Spectrum Authentic
Packaging Glass bottle
Sensitivity Passes Test
Chemical Name or Material Reinecke salt
Show More Show Less
compliance-icons
Product Identifier
  • Reinecke salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'