missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis
Used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
354.00 €
Specifications
CE Marker | Yes |
---|---|
Detectable Analytes | N. meningitidis |
Type | Neisseria Meningitidis Test Kit |
Target Organism Class | Neisseria Meningitidis |
Product Code | Brand | Description | Detectable Analytes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Detectable Analytes | Price | Quantity & Availability | ||||
11414079
|
Thermo Scientific™
R30167001 |
Neisseria meningitidis Group D | N. meningitidis |
354.00 €
2mL |
Estimated Shipment: 01-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Remel Agglutinating Sera are used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
Specifications
Yes | |
N. meningitidis | |
Neisseria Meningitidis Test Kit | |
Neisseria Meningitidis |