Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Proteus O
Remel Agglutinating Sera, Proteus O are used as positive controls for stained Proteus suspensions.
Brand:  Thermo Scientific™ R30165901
Code : NEW
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 15268674
  • 233.00 € / 2mL
Estimated Shipment
12-08-2020
Specifications
Specifications
Proteus OXK
positive controls for stained Proteus suspensions
2mL/Vial
2mL