missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Research™ plus EU-IVD Single Channel Fixed Volume Pipette, Blue
Meets the highest needs in precision and accuracy – combined with ultimate ergonomics and increased flexibility.
Brand: Eppendorf™ 3121000120
Code : 97
Additional Details : Weight : 0.25000kg
Description
- Air-displacement pipette for accurate pipetting of aqueous solutions
- Feel the difference in weight and pipetting forces: the ultra light mechanical pipette is designed according to the strict criteria of the Eppendorf PhysioCare Concept and limits the strain on your hand and arm
- Spring loaded tip cone (not available for 5mL and 10mL pipettes) for minimal tip attachment forces helps to reduce stress
- Low tip ejection force of Eppendorf Research plus can be as low as 3.6 N
- Adjust your pipette in seconds for better accuracy when pipetting various difficult liquids like ethanol or even when pipetting at high altitudes. Return to factory adjustment without calibration
Specifications
±0.6% | |
In vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998 | |
Spring loaded tip cone, ejector, blue operating button | |
1000 μL |
Autoclavable | |
1000μL Pipette tip | |
Blue |