missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Resurge™ CD1 Supplement, without glucose and L-glutamine
A diverse set of fully chemically defined, animal origin–free and protein-free cell culture supplements, which help promote cell growth and boost protein production.
167.00 € - 7660.00 €
Specifications
Content And Storage | Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers. |
---|---|
Cell Line | CHO |
Classification | Animal Origin-free, Chemically-defined, Protein-free |
Culture Type | Mammalian Cell Culture |
Form | Powder |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16279901
|
Gibco™
670011 |
100 g |
167.00 €
100g |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16289901
|
Gibco™
670012 |
1 kg |
1646.00 €
1kg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16299901
|
Gibco™
670013 |
5 kg |
7660.00 €
5kg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
A chemically defined supplement formulation to complement a variety of base media. This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.Specifications
Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers. | |
Animal Origin-free, Chemically-defined, Protein-free | |
Powder | |
No Glutamine | |
Resurge™ |
CHO | |
Mammalian Cell Culture | |
Media Supplement | |
Bottle(s) | |
Animal origin-free |