missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RET [A883F], GST Tag Recombinant Protein
This RET A883F Recombinant Human Protein (658-end) was expressed in insect cells.
524.00 € - 7580.00 €
Specifications
Accession Number | NP_065681.1 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 74.6 kDa |
KinaseFamily | RET Kinase Family |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930413
|
Invitrogen™
A33506 |
10 μg |
524.00 €
10µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17118845
|
Invitrogen™
A33729 |
1 mg |
7580.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17178815
|
Invitrogen™
A33560 |
100 μg |
2435.00 €
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RET receptor tyrosine kinase or ret proto-oncogene is a member of the cadherin superfamily which are cell-surface receptors that transduce signals for cell growth and differentiation. RET can undergo oncogenic activation in vivo and in vitro by cytogenetic rearrangement. Mutations in the RET gene are associated with the disorders multiple endocrine neoplasia, type IIA, multiple endocrine neoplasia, type IIB, Hirschsprung disease, and medullary thyroid carcinoma. RET signaling pathway, by regulating the development of both the nervous and lymphoid system in the gut, plays a key role in the molecular mechanisms that orchestrate intestine organogenesis.- Validated for use with the Z’-LYTE™ assay
- Available in multiple pack sizes for convenient use
Specifications
NP_065681.1 | |
74.6 kDa | |
Human | |
GST Tag | |
Dry Ice | |
Recombinant, Active, Cytoplasmic, Mutant |
Kinase Assay | |
RET Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol | |
Insect | |
Liquid |