missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Knife Mill Grindomix GM300 Accessories: Grinding Containers
Accessories for Knife Mill Grindomix GM300.
457.00 € - 1270.00 €
Specifications
Capacity (Metric) | 5 L |
---|---|
For Use With (Equipment) | For Knife Mill Grindomix GM300 |
Product Code | Brand | Product Type | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Material | Price | Quantity & Availability | ||||
11974302
|
RETSCH
02.045.0056 |
Grinding Container with Knife Holder | Plastic |
900.00 €
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
||||
11984302
|
RETSCH
03.045.0056 |
Grinding Container | Plastic |
457.00 €
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||||
11994302
|
RETSCH
03.045.0055 |
Grinding Container | Stainless Steel |
1270.00 €
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||||
Description
For use with Knife Mill Grindomix GM300 (Cat. No. 08-418-325).
Specifications
5 L | |
For Knife Mill Grindomix GM300 |