missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Test Sieves - 13 Mesh or Less

Stainless steel with wire gauze

Brand:  RETSCH 60.150.050000

Additional Details : Weight : 0.35000kg

 View more versions of this product

Product Code. 10426241

  • 169.00 € / Each
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Dimensions: 8 dia. × 2 in.D (20.3 × 5cm)

Compliance

Meet ASTM E11

Specifications

Specifications

Stainless Steel
8 in.
5.08cm
Test Sieve
8 in.
2 Mesh
Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc.
2 in.
50mm
ISO 3310
Stainless Steel
No. 2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers