missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH ZM200 Ultra Centrifugal Mill: Ring Sieves
Stainless-steel sieves for use with ZM200 Ultra Centrifugal Mill
Brand: RETSCH 03.647.0272
Code : 50
Additional Details : Weight : 0.10000kg
Specifications
Round Hole | |
Stainless Steel | |
3.0mm | |
Ring sieve, With reinforced rim™ |
Reinforced rim | |
0.11 in. | |
ZM200 ultra centrifugal mil |