Learn More
Fisherbrand™ Reusable Class A Volumetric Pipets
Available in either standard or serialized models.
POA
Specifications
Class | A |
---|---|
Material | Borosilicate Glass |
Graduated | Yes |
Graduation Color | Amber |
Tip Style | Tuf-Temp™ treated |
Description
Available in either standard or serialized models.
- Borosilicate glass
- Volume accuracy marked on each pipet
- Permanent amber markings
- Tuf-Temp™–treated tempered tips reduce chipping
Order Info
All pipets are available as PersonaPet™ personalized pipets.
Specifications
A | |
Yes | |
Tuf-Temp™ treated |
Borosilicate Glass | |
Amber | |
Reusable |