Learn More
Thermo Scientific™ RheFlex™ High-Pressure Fittings
Ensure reliable high-pressure connections with Thermo Scientific™ RheFlex™ High-Pressure Fittings.
Brand: Thermo Scientific™ 6000-262
Code : 50
Additional Details : Weight : 0.50000kg
Description
Ensure reliable high-pressure connections by using Thermo Scientific™ RheFlex™ High-Pressure Fittings. These fittings are precision machined from 316 stainless steel to be burr- and contaminant-free.
Specifications
LC Columns | |
Stainless Steel | |
10/Pk. |
10-32, 1/16 in. Extra Long Fittings Set | |
Extra Long High Pressure Fittings Set |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.