missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) octanoate dimer, Thermo Scientific Chemicals

Catalog Number p-7050952
Click to view available options
Quantity:
1 g
73482-96-9
C32H60O8Rh2
778.635
MFCD00064724
FZXFNYFVNKTQSX-UHFFFAOYSA-J
chloro 1,2,3-eta-3-phenyl-2-propenyl 1,3-bis 2,6-di-i-propylphenyl imidazol-2-ylidene palladium ii
131858392
octanoate;rhodium(2+)
CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].[Rh+2].[Rh+2]
73482-96-9
C32H60O8Rh2
778.635
MFCD00064724
FZXFNYFVNKTQSX-UHFFFAOYSA-J
chloro 1,2,3-eta-3-phenyl-2-propenyl 1,3-bis 2,6-di-i-propylphenyl imidazol-2-ylidene palladium ii
131858392
octanoate;rhodium(2+)
CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].[Rh+2].[Rh+2]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Green
Physical Form Powder
Quantity 1 g
Linear Formula [Rh[CH3(CH2)6CO2]2]2
Solubility Information Soluble in hot alcohol, dichloromethane, toluene and acetic acid, slightly soluble in alcohol
Formula Weight 778.63
Chemical Name or Material Rhodium(II) octanoate dimer

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'