Learn More
Brady™ Rigid DuraSleeve™ Labels for M710, M610, M611
Rigid DuraSleeve™ Inserts for use with Brady™ Printers M611, M610 and M710.
Brand:  Brady™ 312099
Additional Details : Weight : 0.06800kg
 View more versions of this product
Product Code. 17847864
  • 42.85 € / Pack of 250
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • Brady™ DuraSleeve™ printable inserts create a durable legend
  • Rigid construction of the DuraSleeve™ wire marking system provides inserts that can easily be inserted into a clean carrier
  • Legend can be changed before or after termination
Specifications
Specifications
Non-adhesive
Gloss
Vinyl
Non-sterile
M610, M710, M611, BMP61, BMP71
250 Labels
-40°C to +70°C
0.249 mm
0.17 in.
30 mm
Yellow
Tape
Wire and Cable Markers
Wire and Cable Labeling
1 Label
-40°F to +158°F
0.0098 in.
4.4 x 30 mm
1.18 in.