missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ROBO2 Pre-design Chimera RNAi
Homo sapiens roundabout, axon guidance receptor, homolog 2 (Drosophila) (ROBO2), transcript variants mRNA. This product targets for all of 5 transcript variants (NM_001128929,NM_001290039,NM_001290040,NM_001290065,NM_002942)
POA
Specifications
Content And Storage | Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. |
---|---|
For Use With (Application) | RNAi Knockdown |
Description
- Gene name: ROBO2
- Gene Alias: KIAA1568,SAX3
- Entrez GeneID: 6092
- Gene Description: Roundabout, axon guidance receptor, homolog 2 (Drosophila)
Specifications
Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. | |
RNAi Knockdown |
For research use only