Learn More
Hettich™ Rotor Adapter for Carrier no. 1752
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
POA
Specifications
Type | Rotor Adapter |
---|---|
For Use With (Equipment) | ROTINA 380/380 R Centrifuge with Carrier no. 1752 |
Description
- For use with ROTINA 380/380 R
- The ROTINA 380's versatility and size make an invaluable addition to any lab. Ideal for clinical, research, food and beverage and industrial applications.
- Robust and steady high-capacity centrifuge
- Accessories with bio-containment protect users from aerosols while ensuring optimal safety, for instance, while centrifuging infectious samples
- Temperature control system with pre-cool function from -20 to +40°C
- Versa-Rotor™ for multiple applications
- All-steel construction
- Quiet Operation
- Easy-Lift™/Safety-Lock™ Lid
Specifications
Rotor Adapter | |
ROTINA 380/380 R Centrifuge with Carrier no. 1752 |