Learn More
Thermo Scientific™ Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Expand individual bucket capacity or maximize total blood bags per spin with choice of buckets for Thermo Scientific™ HAEMAFlex™ blood banking rotors.
Brand:  Thermo Scientific™ 75003836
Additional Details : Weight : 2.73000kg
 View more versions of this product
Product Code. 15880582
  • 1535.00 € / Pack of 2
In Stock
to see stock.
Description
Description
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.
For high-throughput blood processing needs, the HAEMAFlex 12 swinging bucket rotor provides 12 x 500mL capacity or upgrade your centrifuge capacity to 16 x 500mL blood bags with the HAEMAFlex 16 rotor and the same bucket selections.
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Specifications
Specifications
HAEMAFlex 6/8 swinging bucket rotors
16 x 500 mL
Oval, Single Blood Bag Bucket, with Dura-Coat (set of 2)