Learn More
Eppendorf™ Aerosol-Tight Rotor Lids
Providing secure closure for a variety of Eppendorf rotors.
Brand: Eppendorf™ 5490063007
Code : 99
Additional Details : Weight : 0.22500kg
Description
- Aluminim construction
- Aerosol-tight
- Autoclavable for 20 min at 121°C
Specifica
Aerosol-tight Lid | |
Aluminum | |
FA–45-24-11 | |
Autoclavable at 121°C for 20 min. |