missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Rotor S-4
High capacity swing-bucket rotor. Designed for use with Eppendorf 5920 R
3580.00 € - 4305.00 €
Product Code | Brand | Type | Capacity | Max. RCF | Max. Speed | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Capacity | Max. RCF | Max. Speed | Price | Quantity & Availability | ||
15748369
|
Eppendorf™
5895117007 |
Rotor S-4x1000 | 40 × 50 mL | 3,076 x g | 4,250 rpm |
3580.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15768069
|
Eppendorf™
5895118003 |
Rotor S-4x1000 | 8 × 250 mL | 3,153 x g | 3,700 rpm |
4305.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15728379
|
Eppendorf™
5895190006 |
Rotor S-4x Universal-Large | 4 x 750 mL | 4,402 x g | 4,250 rpm |
4220.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Broad range of adapters enables high versatility for tubes and flasks from 0.2mL to 750mL and plates
- Universal bucket design accommodates both plates and tubes
- Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available
- Buckets, caps and adapters are autoclavable
Specifications
Eppendorf 5920R | |
Autoclavable |