missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Rotor S-4x1000
Broad range of adapters enables high versatility for tubes and flasks from 0.2 mL to 1,000 mL and plate. Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available
Brand: Eppendorf™ 5895100007
Additional Details : Weight : 7.92000kg
Description
- Broad range of adapters enables high versatility for tubes and bottles from 0.2mL to 750mL and plates
- Aerodynamic round bucket design for up to 36 % energy savings compared to Rotor A-4-81
- Mixed loading with 2 plate buckets and 2 round tube buckets allows for increased versatility
- Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available
- Rotor life time: max. 100,000 cycles
- Buckets, lids and adapters are autoclavable
Specifications
Rotor S-4x1000, incl. 4 × 1000 mL round buckets | |
3,700 rpm | |
4 x 1,000mL round buckets |
4 x 1000 mL | |
5920R Centrifuge | |
Autoclavable (buckets, caps and adapters) |