missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Round Bottom Polystyrene Test Tubes
Clear, round bottom test tubes for sample transfer.
POA
Specifications
Color | Clear |
---|---|
Material | Polystyrene |
For Use With | Sample transfer |
Sterility | Non-sterile |
Product Type | Test Tube |
Description
Clear, round bottom test tubes for sample transfer.
Polystyrene tubes are ideally suited for optical measurements as a result of its high clarity.
- Nonsterile
- Available in different sizes
Specifications
Clear | |
Sample transfer | |
Test Tube |
Polystyrene | |
Non-sterile |