Learn More
Saint Gobain Life Sciences Rubber Folding Skirted Stopper
Use these stoppers for a variety of applications. Saint Gobain Life Sciences&trade Rubber Folding Skirted Stopper are particularly suited for chemical transfer.
Brand:  Saint Gobain Life Sciences 407007-100
Code : A0
Additional Details : Weight : 0.06000kg
Product Code. 11764588
  • 53.80 € / Pack of 100
In Stock
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Two-part folding skirt stoppers
  • Designed with a double seal to keep container contents secure
  • Ideal for securing glass, metal and plastic containers
  • The lower section grips the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim
  • Upper hollow plug section can be perforated to extract contents without removing the stopper and remains watertight even after several perforations
  • For test tubes
  • Easy to install
Specifications
Specifications
Transparent
7.1mm
Rubber
Rubber Stoppers
100/ck.