Learn More
Saint Gobain Life Sciences Rubber Gas Sample Bag
Use these bags for gas sample analysis. Saint Gobain Life Sciences Rubber Gas Sample Bags feature a cylindrical shape and an ebonite tube and tap.
Brand: Saint Gobain Life Sciences 5607074
Code : A0
Additional Details : Weight : 0.10000kg
Description
- Bags are individually packaged
- Available in packs of ten
- Foil thickness: 5/10mm
- Custom sizes and configurations available on request
Specifications
Gas Sampling Bag | |
2 L | |
Faucet | |
Rubber |