missing translation for 'onlineSavingsMsg'
Learn More

Rubidium, plasma standard solution, Specpure™ Rb 10,000μg/mL, Thermo Scientific Chemicals

Catalog Number p-7049430
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
MFCD00134055
This item is not returnable. View return policy
MFCD00134055

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration Rb2CO3 in 5% HNO3
Quantity 50 mL
UN Number UN3264
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Rubidium plasma standard solution, Rb 10000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'