missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max, Thermo Scientific Chemicals

Catalog Number 15414915
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
7440-18-8
Ru
101.07
MFCD00011207 MFCD03458417
KJTLSVCANCCWHF-UHFFFAOYSA-N
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
23950
CHEBI:30682
ruthenium
[Ru]
This item is not returnable. View return policy
7440-18-8
Ru
101.07
MFCD00011207 MFCD03458417
KJTLSVCANCCWHF-UHFFFAOYSA-N
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
23950
CHEBI:30682
ruthenium
[Ru]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance gray powder
Assay Percent Range >99.99% (Metals basis excluding Ca)
Quantity 1 g
Mesh Size -200 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Ruthenium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'