Learn More
(S)-(+)-1-Amino-2-propanol, 98+%, ACROS Organics™
Brand: Acros Organics 218980010
Code : 29
Additional Details : CAS Number : 2799-17-9 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
2799-17-9 | |
75.11 | |
HXKKHQJGJAFBHI-VKHMYHEASA-N | |
7311736 | |
CC(CN)O |
C3H9NO | |
MFCD00064429 | |
s-+-1-amino-2-propanol, s-1-aminopropan-2-ol, 2s-1-aminopropan-2-ol, +-isopropanolamine, s-1-amino-2-propanol, 2-propanol, 1-amino-, 2s, 1-amino-2-propanol #, 2-propanol, 1-amino-, s, ambotzfal1033 | |
(2S)-1-aminopropan-2-ol |
Specifications
(S)-(+)-1-Amino-2-propanol | |
71°C | |
Glass bottle | |
0.954 | |
160°C | |
19°C to 21°C | |
2799-17-9 | |
100.0 | |
C3H9NO | |
MFCD00064429 | |
s-+-1-amino-2-propanol, s-1-aminopropan-2-ol, 2s-1-aminopropan-2-ol, +-isopropanolamine, s-1-amino-2-propanol, 2-propanol, 1-amino-, 2s, 1-amino-2-propanol #, 2-propanol, 1-amino-, s, ambotzfal1033 | |
HXKKHQJGJAFBHI-VKHMYHEASA-N | |
(2S)-1-aminopropan-2-ol | |
7311736 | |
Liquid After Melting |
0.9540g/mL | |
Authentic | |
1.4465 to 1.4495 | |
+ 18 | |
Colorless to Yellow | |
1g | |
98.0 | |
98% min. (GC) | |
CH3CH(OH)CH2NH2 | |
04, IV, 1665 | |
Solubility in water: soluble. Other solubilities: soluble in organic solvents | |
CC(CN)O | |
75.11 | |
75.11 | |
98+% |