missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Methyl-3-pyrrolidinol, 98%, ee 99%, Thermo Scientific Chemicals

Catalog Number 11946671
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
104641-59-0
C5H11NO
101.15
MFCD03788748
FLVFPAIGVBQGET-UHFFFAOYNA-N
s-+-1-methyl-3-pyrrolidinol, 3s-1-methylpyrrolidin-3-ol, s-1-methyl-3-pyrrolidinol, s-1-methylpyrrolidin-3-ol, 3-pyrrolidinol, 1-methyl-, 3s, s-+-3-hydroxy-n-methylpyrrolidine, s-+-1-methyl-3-hydroxypyrrolidine, s-3-hydroxy-1-methylpyrrolidine, 1-methyl-3-pyrrolidinol, +, s-3-hydroxy-1-methyl-pyrrolidine
643504
CN1CCC(O)C1
This item is not returnable. View return policy
104641-59-0
C5H11NO
101.15
MFCD03788748
FLVFPAIGVBQGET-UHFFFAOYNA-N
s-+-1-methyl-3-pyrrolidinol, 3s-1-methylpyrrolidin-3-ol, s-1-methyl-3-pyrrolidinol, s-1-methylpyrrolidin-3-ol, 3-pyrrolidinol, 1-methyl-, 3s, s-+-3-hydroxy-n-methylpyrrolidine, s-+-1-methyl-3-hydroxypyrrolidine, s-3-hydroxy-1-methylpyrrolidine, 1-methyl-3-pyrrolidinol, +, s-3-hydroxy-1-methyl-pyrrolidine
643504
CN1CCC(O)C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Brown to Colorless
Density 0.9930g/mL
Boiling Point 181°C to 182°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 1 g
Specific Gravity 0.993
Specific Rotation 7
IUPAC Name (3S)-1-methylpyrrolidin-3-ol
Formula Weight 101.15
Percent Purity 98%,ee99%
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-Methyl-3-pyrrolidinol, ee 99%
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-1-Methyl-3-pyrrolidinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'