missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride, 95%, 98% ee, ACROS Organics™

Brand:  Acros Organics 362040250

Code : 29

Additional Details : CAS Number : 167834-24-4 Weight : 0.07500kg

Quantity 25g

 View more versions of this product

Product Code. 11955321

  • 2108.00 € / 25g

Estimated Shipment: 08-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

167834-24-4
194.25
NUWRDXMXYDWUAN-JTQLQIEISA-O
22831526
CCOC(=O)C[C@H]([NH3+])C1=CC=CC=C1
C11H16NO2
MFCD02259715
s-ethyl 3-amino-3-phenylpropanoate hydrochloride, s-3-amino-3-phenylpropanoic acid ethyl ester hydrochloride, ethyl 3s-3-amino-3-phenylpropanoate hydrochloride, 3s-3-amino-3-phenylpropanoic acid ethyl ester hcl, s-beta-phenylalanine ethyl ester hydrochloride, pubchem13852, s-b-phenylalanine ethyl ester hcl, ethyl s-beta-aminobenzenepropanoate hcl, s-3-amino-3-phenylpropanoicacidethylesterhcl, ethyl 3s-3-amino-3-phenylpropanoate, chloride
ethyl (3S)-3-amino-3-phenylpropanoate;hydrochloride

Specifications

(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride
Authentic
White
25g
94% min. (HPLC)
MFCD02259715
NUWRDXMXYDWUAN-JTQLQIEISA-O
ethyl (3S)-3-amino-3-phenylpropanoate;hydrochloride
22831526
Powder
97.5% (GC) min.
Glass Bottle
117.0°C
167834-24-4
C11H16NO2
s-ethyl 3-amino-3-phenylpropanoate hydrochloride, s-3-amino-3-phenylpropanoic acid ethyl ester hydrochloride, ethyl 3s-3-amino-3-phenylpropanoate hydrochloride, 3s-3-amino-3-phenylpropanoic acid ethyl ester hcl, s-beta-phenylalanine ethyl ester hydrochloride, pubchem13852, s-b-phenylalanine ethyl ester hcl, ethyl s-beta-aminobenzenepropanoate hcl, s-3-amino-3-phenylpropanoicacidethylesterhcl, ethyl 3s-3-amino-3-phenylpropanoate, chloride
CCOC(=O)C[C@H]([NH3+])C1=CC=CC=C1
194.25
229.71
95%,98% ee
Videos
SDS Documents
Certificates