missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride, 95%, 98% ee, Thermo Scientific Chemicals

Catalog Number p-4510672
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
167834-24-4
C11H16NO2
194.25
MFCD02259715
NUWRDXMXYDWUAN-JTQLQIEISA-O
s-ethyl 3-amino-3-phenylpropanoate hydrochloride, s-3-amino-3-phenylpropanoic acid ethyl ester hydrochloride, ethyl 3s-3-amino-3-phenylpropanoate hydrochloride, 3s-3-amino-3-phenylpropanoic acid ethyl ester hcl, s-beta-phenylalanine ethyl ester hydrochloride, pubchem13852, s-b-phenylalanine ethyl ester hcl, ethyl s-beta-aminobenzenepropanoate hcl, s-3-amino-3-phenylpropanoicacidethylesterhcl, ethyl 3s-3-amino-3-phenylpropanoate, chloride
22831526
ethyl (3S)-3-amino-3-phenylpropanoate;hydrochloride
CCOC(=O)C[C@H]([NH3+])C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
167834-24-4
C11H16NO2
194.25
MFCD02259715
NUWRDXMXYDWUAN-JTQLQIEISA-O
s-ethyl 3-amino-3-phenylpropanoate hydrochloride, s-3-amino-3-phenylpropanoic acid ethyl ester hydrochloride, ethyl 3s-3-amino-3-phenylpropanoate hydrochloride, 3s-3-amino-3-phenylpropanoic acid ethyl ester hcl, s-beta-phenylalanine ethyl ester hydrochloride, pubchem13852, s-b-phenylalanine ethyl ester hcl, ethyl s-beta-aminobenzenepropanoate hcl, s-3-amino-3-phenylpropanoicacidethylesterhcl, ethyl 3s-3-amino-3-phenylpropanoate, chloride
22831526
ethyl (3S)-3-amino-3-phenylpropanoate;hydrochloride
CCOC(=O)C[C@H]([NH3+])C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 117.0°C
Color White
Assay Percent Range 94% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Formula Weight 229.71
Percent Purity 95%,98% ee
Quantity 1 g
Physical Form Powder
Chemical Name or Material (S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'