Learn More
(S)-(-)-3-(Dimethylamino)pyrrolidine, 97%, ACROS Organics™
Brand: Acros Organics 426450010
Code : 29
Additional Details : CAS Number : 132883-44-4 Weight : 0.05100kg Transport : UN number : 2733 Chem class : 3 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
132883-44-4 | |
114.19 | |
3s---3-dimethylamino pyrrolidine, s-3-dimethylaminopyrrolidine, s---3-dimethylamino pyrrolidine, s-n,n-dimethylpyrrolidin-3-amine, 3s-n,n-dimethylpyrrolidin-3-amine, s-3-dimethylamino pyrrolidine, 3s-n,n-dimethyl-3-pyrrolidinamine, 3s-3-dimethylaminopyrrolidine, 3-pyrrolidinamine, n,n-dimethyl-, 3s, s-dimethyl-pyrrolidin-3-yl-amine | |
(3S)-N,N-dimethylpyrrolidin-3-amine |
C6H14N2 | |
AVAWMINJNRAQFS-LURJTMIESA-N | |
7019156 | |
CN(C)C1CCNC1 |
Specifications
(S)-(-)-3-(Dimethylamino)pyrrolidine | |
51°C | |
0.899 | |
1g | |
96% (GC) | |
3s---3-dimethylamino pyrrolidine, s-3-dimethylaminopyrrolidine, s---3-dimethylamino pyrrolidine, s-n,n-dimethylpyrrolidin-3-amine, 3s-n,n-dimethylpyrrolidin-3-amine, s-3-dimethylamino pyrrolidine, 3s-n,n-dimethyl-3-pyrrolidinamine, 3s-3-dimethylaminopyrrolidine, 3-pyrrolidinamine, n,n-dimethyl-, 3s, s-dimethyl-pyrrolidin-3-yl-amine | |
CN(C)C1CCNC1 | |
114.19 | |
114.19 |
0.8990g/mL | |
Glass bottle | |
164.0°C | |
132883-44-4 | |
C6H14N2 | |
AVAWMINJNRAQFS-LURJTMIESA-N | |
(3S)-N,N-dimethylpyrrolidin-3-amine | |
7019156 | |
97% |
Safety and Handling
- (S)-(-)-3-(Dimethylamino)pyrrolidine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.