missing translation for 'onlineSavingsMsg'
Learn More

S-Butyrylthiocholine iodide, 98%, Thermo Scientific Chemicals

Catalog Number 11955721
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1866-16-6
C9H20INOS
317.22
WEQAAFZDJROSBF-UHFFFAOYSA-M
butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
74630
2-butanoylsulfanylethyl(trimethyl)azanium;iodide
CCCC(=O)SCC[N+](C)(C)C.[I-]
This item is not returnable. View return policy
1866-16-6
C9H20INOS
317.22
WEQAAFZDJROSBF-UHFFFAOYSA-M
butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
74630
2-butanoylsulfanylethyl(trimethyl)azanium;iodide
CCCC(=O)SCC[N+](C)(C)C.[I-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 172.0°C to 176.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CH2CH2COSCH2CH2N(CH3)3I
Quantity 100 g
Beilstein 04,IV,1585
Solubility Information Solubility in water: freely soluble
Formula Weight 317.22
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material S-Butyrylthiocholine iodide
Show More Show Less
compliance-icons
Product Identifier
  • S-Butyrylthiocholine iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'