missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Glycidyl nosylate, 98%, ACROS Organics™

Brand:  Acros Organics 296420500

Code : 29

Additional Details : CAS Number : 115314-14-2 Weight : 0.10000kg


Packaging Glass bottle
Quantity 50g

 View more versions of this product

Product Code. 11975151

  • 675.00 € / 50g

Estimated Shipment
28-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

115314-14-2
259.23
s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate
[(2S)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate
C9H9NO6S
AIHIHVZYAAMDPM-QMMMGPOBSA-N
146490
C1C(O1)COS(=O)(=O)C2=CC=CC(=C2)[N+](=O)[O-]

Specifications

(S)-(+)-Glycidyl nosylate
Glass bottle
+ 21.50
61.0°C to 65.0°C
115314-14-2
C9H9NO6S
AIHIHVZYAAMDPM-QMMMGPOBSA-N
[(2S)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate
146490
Needles or Powder
Authentic
+ 21.50 (20.00°C c=2,CHCl3)
Brown-Yellow to White
50g
97.5% min. (GC)
s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate
C1C(O1)COS(=O)(=O)C2=CC=CC(=C2)[N+](=O)[O-]
259.23
259.23
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (S)-(+)-Glycidyl nosylate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents