missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-N-FMOC-alpha-Methylphenylalanine sesquihydrate, 98%, 98% ee, Thermo Scientific™
Click to view available options
Quantity:
100 mg
Specifications
CAS | 135944-05-7 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | White to Yellow |
Enantiomeric Excess | 98.0% min. |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Molecular Formula | 1·5 H2O |
Quantity | 100 mg |
Specific Rotation | − 29.00 |
Show More |
Safety and Handling
Product Identifier
- (S)-N-FMOC-alpha-Methylphenylalanine sesquihydrate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'