Learn More
Crowthorne Hi-Tech Services Safety cabinet HLF18 horizontal laminar flow recirculating with 0.3μm Hepa filter bench top 230V 50/60Hz 200W 1880mm x 740mm x 1290mm w x d x h airflow 0.35-0.55 m/sec Labcaire
Brand:  Crowthorne Hi-Tech Services LA-HLF18
Safety cabinet HLF18 horizontal laminar flow recirculating with 0.3μm Hepa filter bench top 230V 50/60Hz 200W 1880mm x 740mm x 1290mm w x d x h airflow 0.35-0.55 m/sec Labcaire
Code : 04
Additional Details : Weight : 70.00000kg
Product Code. 12791476