Learn More
Crowthorne Hi-Tech Services Safety cabinet VLF18UV vertical laminar flow recirculating with 0.3μm Hepa filter 240V 50/60Hz 200W 1930mm x 730mm x 1340mm w x d x h airflow 0.35-0.55 m/sec Labcaire
Brand:  Crowthorne Hi-Tech Services LA-VLF18UV
Safety cabinet VLF18UV vertical laminar flow recirculating with 0.3μm Hepa filter 240V 50/60Hz 200W 1930mm x 730mm x 1340mm w x d x h airflow 0.35-0.55 m/sec Labcaire
Code : 04
Additional Details : Weight : 70.00000kg
Product Code. 12797876