Learn More
Crowthorne Hi-Tech Services Fume cabinet T400 mobile filtration recirculating 20kg ED type activated carbon filter free standing 240V 50/60Hz 200W airflow >0.5 m/sec Labcaire
Brand: Crowthorne Hi-Tech Services LA-T400L
Fume cabinet T400 mobile filtration recirculating 20kg ED type activated carbon filter free standing 240V 50/60Hz 200W airflow >0.5 m/sec Labcaire
Code : 04
Additional Details : Weight : 200.00000kg