missing translation for 'onlineSavingsMsg'
Learn More

Sand, Extra Pure, SLR, Ottawa, 20-30 Mesh, Fisher Chemical™

Brand:  Fisher Chemical S/0365/63

95.00 EUR valid until 2021-08-31
Use promo code "14805" to get your promotional price.



Code : 21

Additional Details : CAS Number : 14808-60-7 Weight : 3.00000kg

Quantity 3kg

 View more versions of this product

Product Code. 10020262

  • 100.00 € / 3kg

Estimated Shipment: 10-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sand, Extra Pure, SLR, Ottawa, 20-30 Mesh

Specifications

Chemical Identifiers

14808-60-7
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
O=[Si]=O

Specifications

14808-60-7
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.08g/mol
Plastic powder jar
Beige
3kg
O2Si
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
O=[Si]=O
60.08
CHEBI:30563
Solid
2230°C
1610°C
Videos
SDS Documents
Certificates