Learn More
Sartorius Qualitative Grade 292 a Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Brand:  Sartorius FT-3-215-110
Code : NEW
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10062401
  • 16.10 € / Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
Disc
97 sec.
100/Pk.
110 mm
4 to 7 μm
292 a
0.19 mm