missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Qualitative Grade 292 a Filter Papers

These filter papers are especially suitable for qualitative analyses that require a low ash content.

Brand:  Sartorius FT-3-215-240

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10260672

  • 65.50 € / Pack of 100
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications

Specifications

Disc
97 sec.
100/Pk.
240 mm
4 to 7 μm
292 a
0.19 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers