Learn More
Sartorius Qualitative Grade 292 a Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Brand:  Sartorius FT-3-215-320
Additional Details : Weight : 0.81000kg
 View more versions of this product
Product Code. 15992900
  • 69.30 € / Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
Disc
97 sec.
100/Pk.
320 mm
4 to 7 μm
292 a
0.18 mm