missing translation for 'onlineSavingsMsg'
Learn More

Sartorius MGD Grade Glass Microfiber Filters w/o Binder

Binder free glass microfiber filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.

Brand:  Sartorius FT-3-1104-240

Additional Details : Weight : 0.27100kg

Product Code. 15902870

  • 231.00 € / Pack of 25
Estimated Shipment: 27-05-2024
to see stock.
Add to basket

Description

Description

Binder free glass microfiber filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.
Specifications

Specifications

18.5mbar
Disc
2.7 μm
18 sec.
MGD
500°C
0.53 mm
24 cm
White
Borosilicate Glass Microfiber
Non-sterile
Ideal for critical analyses in which impurities can be washed out of the filter
932°F
0.02 in.
9.4 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers