Learn More
Sartorius Quantitative Grade 390 Filter Papers
These filter papers are used for quantitative and gravimetric analyses as well as for pressure or vacuum filtration. They are made out of 100 % cotton linters with an α-cellulose content of > 98 % and are acid-washed to make the papers ashless and achieve high purity.
12.30 € - 276.00 €
Product Code | Brand | Format | Filtration Speed | Particle Retention | Diameter (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Filtration Speed | Particle Retention | Diameter (Metric) | Price | Quantity & Availability | ||
10016861
|
Sartorius
FT-3-103-110 |
Disc | 100 sec. | 3 to 5 μm | 110 mm |
35.50 €
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
10541922
|
Sartorius
FT-3-103-125 |
Disc | 100 sec. | 3 to 5 μm | 125 mm |
41.70 €
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
10042431
|
Sartorius
FT-3-103-185 |
Disc | 100 sec. | 3 to 5 μm | 185 mm |
89.60 €
Pack of 100 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||
10551922
|
Sartorius
FT-3-103-150 |
Disc | 100 sec. | 3 to 5 μm | 150 mm |
59.60 €
Pack of 100 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||
11711385
|
Sartorius
FT-4-103-150 |
Folded | 100 sec. | 3 to 5 μm | 125 mm |
83.00 €
Pack of 100 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||
10575891
|
Sartorius
FT-3-103-055 |
Disc | 100 sec. | 3 to 5 μm | 55 mm |
12.30 €
Pack of 100 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||
10092401
|
Sartorius
FT-3-103-090 |
Disc | 100 sec. | 3 to 5 μm | 90 mm |
25.80 €
Pack of 100 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
10624522
|
Sartorius
FT-3-103-070 |
Disc | 100 sec. | 3 to 5 μm | 70 mm |
16.10 €
Pack of 100 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
11701385
|
Sartorius
FT-4-103-125 |
Folded | 250 sec. | 1 to 2 μm | 125 mm |
69.30 €
Pack of 100 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||
15932980
|
Sartorius
FT-4-103-185 |
Folded | 100 sec. | 3 to 5 μm | 185 mm |
102.00 €
Pack of 100 |
Estimated Shipment: 23-04-2024
Log in to see stock. |
||
10576081
|
Sartorius
FT-3-103-240 |
Disc | 100 sec. | 3 to 5 μm | 240 mm |
276.00 €
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
Description
- Ash-free (ash content ≤ 0.01 % according to DIN 540370)
- Wet-strengthened
- Color-coded box for easy selection
Specifications
Green | |
390 | |
100/Pk. |