missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Sartolab™ RF/BT Vacuum Filtration Units
Filter aqueous solutions quickly and easily. Sartorius Sartolab™ RF/BT Vacuum Filtration Units are disposable and easy to use filter units. They contain a high quality PES membrane and combine the highest flow-rates and throughput with extremely low protein binding and extractables. Available in a range of volumes.
119.00 € - 342.00 €
Specifications
Certifications/Compliance | USP Class VI, ISO 10993 |
---|---|
Connectors | Plug-on hose nipple |
Disposable | Single-use |
For Use With (Equipment) | 1.77 in. bottle neck |
Material | Polystyrene |
Product Code | Brand | Type | Filtration Area | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Filtration Area | Volume (Metric) | Price | Quantity & Availability | |||
11771005
|
Sartorius
180C1----------E |
Sartolab RF Filter | 18 cm2 | 150 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11781005
|
Sartorius
180C2----------E |
Sartolab RF Filter | 39 cm2 | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11711015
|
Sartorius
180C6----------E |
Sartolab BT | 62 cm2 | 1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11755815
|
Sartorius
180C4----------K |
Sartolab BT | 18 cm2 | 150 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11721015
|
Sartorius
180C7----------E |
Sartolab RF Filter | 24 cm2 | 250 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11701015
|
Sartorius
180C5----------E |
Sartolab BT | 39 cm2 | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11791005
|
Sartorius
180C3----------E |
Sartolab RF Filter | 62 cm2 | 1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Quality, reliable sterile filtration units- Receiver Flask (RF) and Bottle Top (BT) units contain a high quality Sartorius PES membrane
- Combine the highest flow-rates and throughput with extremely low protein binding and extractables
- Ideal for filtration of tissue culture media, buffers, additives and other aqueous biologicals
- RF and BT vacuum units are available as 150mL, 250mL, 500mL and 1000mL units
- 500mL and 1000mL units come standard with 90mm membranes
- Improved filtration speed and throughput for cost effective filtration
Specifications
USP Class VI, ISO 10993 | |
Single-use | |
Polystyrene | |
0.22 μm | |
Filtration of tissue culture media, buffers, additives and other aqueous biologicals |
Plug-on hose nipple | |
1.77 in. bottle neck | |
PES | |
Sterile |