Learn More
Sartorius Reusable Sterility Testing System: Silicone Adapter
Silicone Adapter for use with Reusable Sterility Testing System.
Brand:  Sartorius 16968
Additional Details : Weight : 0.03400kg
Product Code. 15922560
  • 25.30 € / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Description
Description
  • With stoppers drilled to fit filter holder base
  • Allows sterile filtration system to be used with Sartorius and other stainless-steel and PVC manifolds
Specifications
Specifications
Reusable
Silicone
Silicone Adapter
Reusable Sterility Testing System
Non-sterile